Assay Details
| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013973.1 | NP_001013995.1 | 10 - 11 | 2522 | 73 | |
| XM_006247579.2 | XP_006247641.1 | 10 - 11 | 2690 | 73 | ||
| XM_006247580.2 | XP_006247642.1 | 11 - 12 | 2809 | 73 | ||
| GenBank mRNA | AB552845.1 | - | 8 - 9 | 2267 | 73 | |
| BC083567.1 | - | 10 - 11 | 2522 | 7112851 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

March10
Genomic Map