Assay Details
| Gene Symbol: Fam58b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025412.1 | NP_001020583.1 | 1 | 895 | 88 | |
| GenBank mRNA | BC086445.1 | - | 1 | 882 | 88 | |
| BC097941.1 | - | 1 | 895 | 7376750 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam58b
Genomic Map