Assay Details
| Gene Symbol: Mrpl45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105834.1 | NP_001099304.1 | 3 - 4 | 406 | 156 | |
| GenBank mRNA | FQ216878.1 | - | 2 - 3 | 198 | 156 | |
| FQ225351.1 | - | 2 - 3 | 200 | 156 | ||
| FQ232005.1 | - | 2 - 3 | 198 | 156 | ||
| FQ233183.1 | - | 2 - 3 | 198 | 156 | ||
| FQ233508.1 | - | 2 - 3 | 198 | 156 | ||
| FQ233844.1 | - | 2 - 3 | 156 | |||
| FQ234322.1 | - | 2 - 3 | 198 | 156 | ||
| FQ234954.1 | - | 2 - 3 | 198 | 156 | ||
| FQ235000.1 | - | 2 - 3 | 198 | 156 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl45
Genomic Map