Assay Details
| Gene Symbol: Mrpl45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105834.1 | NP_001099304.1 | 6 - 7 | 708 | 93 | |
| GenBank mRNA | FQ216878.1 | - | 5 - 6 | 500 | 93 | |
| FQ225351.1 | - | 5 - 6 | 502 | 93 | ||
| FQ226279.1 | - | 5 - 6 | 500 | 93 | ||
| FQ229232.1 | - | 5 - 6 | 498 | 93 | ||
| FQ232005.1 | - | 5 - 6 | 501 | 94 | ||
| FQ233183.1 | - | 5 - 6 | 500 | 93 | ||
| FQ233508.1 | - | 5 - 6 | 500 | 93 | ||
| FQ234954.1 | - | 5 - 6 | 500 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl45
Genomic Map