Assay Details
| Gene Symbol: Med7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286183.1 | NP_001273112.1 | 1 - 2 | 47 | 92 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286183.1 transcripts.
Set Membership: |
|

Med7
Genomic Map