Assay Details
| Gene Symbol: Map3k14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108301.1 | NP_001101771.1 | 6 - 7 | 1385 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
protein phosphorylation
immune response
I-kappaB kinase/NF-kappaB signaling
signal transduction by protein phosphorylation
intracellular signal transduction
NIK/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to mechanical stimulus
protein kinase activity
receptor signaling protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
MAP kinase kinase activity
ATP binding
MAPK cascade
activation of MAPK activity
protein phosphorylation
immune response
I-kappaB kinase/NF-kappaB signaling
signal transduction by protein phosphorylation
intracellular signal transduction
NIK/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to mechanical stimulus
protein kinase activity
receptor signaling protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
MAP kinase kinase activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P091620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156376 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091618 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091618 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P091618 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P497290 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75797 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20180 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10748026 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748035 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748027 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748022 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748032 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748034 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748024 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748029 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748030 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748031 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748025 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748036 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748041 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748033 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748028 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10748037 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10748021 | - | - |
| Rat Genome 230 2.0 Array | 1391688_at | - | - |
| Rat Genome 230 2.0 Array | 1391688_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391688_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391688_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000035280.rn.1 | TC1000003898.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391688_at | - | - |
| Rat Expression Set 230 | 1391688_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748021 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748037 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20973016 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359019 | - | - |

Map3k14
Genomic Map