Assay Details
| Gene Symbol: Hn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013182.1 | NP_001013200.1 | 2 - 3 | 199 | 98 | |
| XM_006245994.2 | XP_006246056.1 | 2 - 3 | 613 | 98 | ||
| GenBank mRNA | BC091238.1 | - | 2 - 3 | 199 | 7323726 | 98 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hn1l
Genomic Map