Assay Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108297.1 | NP_001101767.1 | 10 - 11 | 1116 | 60 | |
| XM_006247347.1 | XP_006247409.1 | 10 - 11 | 1151 | 60 | ||
| GenBank mRNA | BC112385.1 | - | 9 - 10 | 1986 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsdma
Genomic Map