Assay Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108297.1 | NP_001101767.1 | 3 - 4 | 494 | 70 | |
| XM_006247347.1 | XP_006247409.1 | 3 - 4 | 529 | 70 | ||
| GenBank mRNA | BC112385.1 | - | 3 - 4 | 478 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsdma
Genomic Map