Assay Details
| Gene Symbol: LOC303566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017988.1 | NP_001017988.1 | 7 - 8 | 1837 | 97 | |
| XM_006247329.2 | XP_006247391.1 | 8 - 9 | 1910 | 97 | ||
| GenBank mRNA | AY623029.1 | - | 4 - 5 | 388 | 97 | |
| AY623030.1 | - | 7 - 8 | 1837 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC303566
Genomic Map