Assay Details
| Gene Symbol: Dusp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247402.2 | XP_006247464.1 | 2 - 3 | 278 | 63 | |
| XM_006247403.2 | XP_006247465.1 | 2 - 3 | 277 | 63 | ||
| GenBank mRNA | BC169043.1 | - | 1 - 2 | 147 | 9099479 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Dusp3
Genomic Map