Assay Details
| Gene Symbol: Ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037643.2 | NP_001032732.2 | 2 - 3 | 430 | 68 | |
| GenBank mRNA | BC107663.1 | - | 2 - 3 | 430 | 7936709 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2z
Genomic Map