Assay Details
| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105773.1 | NP_001099243.1 | 15 - 16 | 2001 | 56 | |
| XM_006245936.2 | XP_006245998.1 | 15 - 16 | 2039 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chtf18
Genomic Map