Assay Details
| Gene Symbol: Mgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246183.2 | XP_006246245.1 | 1 - 2 | 686 | 81 | |
| GenBank mRNA | AB012876.1 | - | 1 - 2 | 210 | 81 | |
| BC074010.1 | - | 1 - 2 | 58 | 7100030 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC074010.1, XM_006246183.2 transcripts.
Set Membership: |
|

Mgat1
Genomic Map