Assay Details
| Gene Symbol: Sgsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107020.1 | NP_001100490.1 | 12 - 13 | 1629 | 89 | |
| XM_006246740.1 | XP_006246802.1 | 13 - 14 | 1987 | 89 | ||
| XM_006246741.2 | XP_006246803.1 | 12 - 13 | 2295 | 89 | ||
| XM_008768009.1 | XP_008766231.1 | 13 - 14 | 1987 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm2
Genomic Map