Assay Details
| Gene Symbol: Dnase1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001057412.3 | XP_001057412.4 | 2 - 3 | 438 | 66 | |
| XM_006246041.1 | XP_006246103.1 | 2 - 3 | 438 | 66 | ||
| XM_006246042.1 | XP_006246104.1 | 2 - 3 | 438 | 66 | ||
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246025.2 | XP_006246087.1 | 14 - 15 | 4917 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Dnase1l2
Genomic Map