Assay Details
| Gene Symbol: LOC100911674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750836.3 | XP_003750884.1 | 2 - 3 | 971 | 85 | |
| Gene Symbol: Natd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170541.1 | NP_001164012.1 | 2 - 3 | 241 | 85 | |
| GenBank mRNA | CK480689.1 | - | 2 - 3 | 241 | 85 | |
| FQ231549.1 | - | 2 - 3 | 283 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911674
Genomic Map