Assay Details
| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767435.1 | XP_008765657.1 | 5 - 6 | 1870 | 81 | |
| XM_008767436.1 | XP_008765658.1 | 5 - 6 | 538 | 81 | ||
More Information
Set Membership: |
|

L3mbtl4
Genomic Map