Assay Details
| Gene Symbol: Stx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012151.1 | NP_001012151.1 | 10 - 11 | 925 | 96 | |
| GenBank mRNA | BC079183.1 | - | 10 - 11 | 925 | 7130966 | 96 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
membrane fusion
endoplasmic reticulum membrane organization
positive regulation of organelle assembly
positive regulation of ER to Golgi vesicle-mediated transport
regulation of Golgi organization
SNAP receptor activity
protein binding
protein domain specific binding
intracellular protein transport
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
membrane fusion
endoplasmic reticulum membrane organization
positive regulation of organelle assembly
positive regulation of ER to Golgi vesicle-mediated transport
regulation of Golgi organization
SNAP receptor activity
protein binding
protein domain specific binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P410000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100014 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P410000 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P410000 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P410000 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P410000 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38246 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50054 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41490 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88144 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57872 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78666 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68289 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41441 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10777345 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777343 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777338 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777341 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777342 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777339 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777344 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777346 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777340 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777349 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10777337 | - | - |
| Rat Genome 230 2.0 Array | 1372512_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372512_at | - | - |
| Affymetrix HT Rat Focus Array | 1372512_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372512_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400014364.rn.1 | TC1400002025.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372512_at | - | - |
| Rat Expression Set 230 | 1372512_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777337 | - | - |
| Rat Genome U34 Set | rc_AI638998_at | - | - |
| Rat Genome U34 Set | rc_AI172102_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21371790 | - | - |

Stx18
Genomic Map