Assay Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270871.1 | NP_001257800.1 | 13 - 14 | 1093 | 102 | |
| NM_001270872.1 | NP_001257801.1 | 12 - 13 | 1057 | 102 | ||
| NM_181366.2 | NP_852031.1 | 14 - 15 | 1141 | 102 | ||
| XM_006256916.2 | XP_006256978.1 | 14 - 15 | 1181 | 102 | ||
| XM_008773154.1 | XP_008771376.1 | 13 - 14 | 1521 | 102 | ||
| XM_008773155.1 | XP_008771377.1 | 13 - 14 | 1131 | 102 | ||
| XM_008773156.1 | XP_008771378.1 | 12 - 13 | 1485 | 102 | ||
| XM_008773157.1 | XP_008771379.1 | 12 - 13 | 1479 | 102 | ||
| XM_008773158.1 | XP_008771380.1 | 11 - 12 | 1443 | 102 | ||
| XM_008773159.1 | XP_008771381.1 | 10 - 11 | 1034 | 102 | ||
| XM_008773160.1 | XP_008771382.1 | 13 - 14 | 1522 | 102 | ||
| GenBank mRNA | AF538953.1 | - | 14 - 15 | 1151 | 102 | |
| AF538958.1 | - | 13 - 14 | 1103 | 102 | ||
| AF538959.1 | - | 12 - 13 | 1067 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr64
Genomic Map