Assay Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270871.1 | NP_001257800.1 | 21 - 22 | 2213 | 135 | |
| NM_001270872.1 | NP_001257801.1 | 20 - 21 | 2177 | 135 | ||
| NM_181366.2 | NP_852031.1 | 22 - 23 | 2261 | 135 | ||
| XM_006256916.2 | XP_006256978.1 | 22 - 23 | 2301 | 135 | ||
| XM_008773154.1 | XP_008771376.1 | 21 - 22 | 2641 | 135 | ||
| XM_008773155.1 | XP_008771377.1 | 21 - 22 | 2251 | 135 | ||
| XM_008773156.1 | XP_008771378.1 | 20 - 21 | 2605 | 135 | ||
| XM_008773157.1 | XP_008771379.1 | 20 - 21 | 2599 | 135 | ||
| XM_008773158.1 | XP_008771380.1 | 19 - 20 | 2563 | 135 | ||
| XM_008773159.1 | XP_008771381.1 | 18 - 19 | 2154 | 135 | ||
| XM_008773160.1 | XP_008771382.1 | 21 - 22 | 2642 | 135 | ||
| GenBank mRNA | AF538953.1 | - | 22 - 23 | 2271 | 135 | |
| AF538958.1 | - | 21 - 22 | 2223 | 135 | ||
| AF538959.1 | - | 20 - 21 | 2187 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr64
Genomic Map