Assay Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270872.1 | NP_001257801.1 | 3 - 4 | 205 | 92 | |
| XM_008773156.1 | XP_008771378.1 | 3 - 4 | 633 | 92 | ||
| GenBank mRNA | AF538959.1 | - | 3 - 4 | 215 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr64
Genomic Map