Assay Details
| Gene Symbol: Eif1ax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106963.1 | NP_001100433.1 | 3 - 4 | 408 | 108 | |
| GenBank mRNA | BC168938.1 | - | 3 - 4 | 281 | 7097069 | 108 |
| FQ217925.1 | - | 3 - 4 | 373 | 108 | ||
| FQ221854.1 | - | 3 - 4 | 387 | 108 | ||
| FQ222873.1 | - | 3 - 4 | 406 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif1ax
Genomic Map