Assay Details
| Gene Symbol: LOC103693605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768923.1 | XP_008767145.1 | 2 - 3 | 1891 | 64 | |
| XM_008768925.1 | XP_008767147.1 | 2 - 3 | 1891 | 64 | ||
| XM_008768926.1 | XP_008767148.1 | 2 - 3 | 1891 | 64 | ||
| Gene Symbol: Car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105901.1 | NP_001099371.1 | 2 - 3 | 173 | 64 | |
| XM_008768868.1 | XP_008767090.1 | 2 - 3 | 1891 | 64 | ||
| XM_008768869.1 | XP_008767091.1 | 2 - 3 | 1891 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103693605
Genomic Map