Assay Details
| Gene Symbol: LOC103693605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768923.1 | XP_008767145.1 | 5 - 6 | 2318 | 84 | |
| XM_008768924.1 | XP_008767146.1 | 4 - 5 | 2282 | 84 | ||
| XM_008768926.1 | XP_008767148.1 | 5 - 6 | 2318 | 84 | ||
| Gene Symbol: Car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105901.1 | NP_001099371.1 | 5 - 6 | 600 | 84 | |
| XM_008768867.1 | XP_008767089.1 | 4 - 5 | 2282 | 84 | ||
| XM_008768869.1 | XP_008767091.1 | 5 - 6 | 2318 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103693605
Genomic Map