Assay Details
| Gene Symbol: Klhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106054.1 | NP_001099524.1 | 1 - 2 | 1176 | 68 | |
| XM_008770864.1 | XP_008769086.1 | 1 - 2 | 1332 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl1
Genomic Map