Assay Details
| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017505.1 | NP_001017505.1 | 8 - 9 | 1220 | 82 | |
| GenBank mRNA | BC087076.1 | - | 8 - 9 | 1220 | 7134947 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P106573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76603 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P106574 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P106574 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P106574 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78271 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88583 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38641 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10620 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19035 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21061 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75713 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40615 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23349 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64847 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2387 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76128 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66857 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47529 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55998 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77163 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27981 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100601 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24669 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10782198 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782193 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782191 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782192 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782196 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782195 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782197 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782194 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782188 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10782187 | - | - |
| Rat Genome 230 2.0 Array | 1383708_at | - | - |
| Rat Genome 230 2.0 Array | 1383708_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383708_at | - | - |
| Affymetrix HT Rat Focus Array | 1383708_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500007631.rn.1 | TC1500001218.rn.1 | RTA TC |
| Rat Expression Set 230 | 1383708_at | - | - |
| Rat Expression Set 230 | 1383708_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782187 | - | - |
| Rat Genome U34 Set | rc_AI235334_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21618568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650212 | - | - |

Itgbl1
Genomic Map