Assay Details
| Gene Symbol: Rad21l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001072159.4 | XP_001072159.3 | 9 - 10 | 1260 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad21l1| Gene Symbol: Rad21l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001072159.4 | XP_001072159.3 | 9 - 10 | 1260 | 75 | |
Set Membership: |
|