Assay Details
| Gene Symbol: Kif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106529.1 | NP_001099999.1 | 2 - 3 | 1542 | 63 | |
| XM_006235287.2 | XP_006235349.1 | 2 - 3 | 1963 | 63 | ||
| XM_006235288.1 | XP_006235350.1 | 2 - 3 | 1579 | 63 | ||
| XM_006235289.2 | XP_006235351.1 | 2 - 3 | 1759 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif3b
Genomic Map