Assay Details
Gene Symbol: Dapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108582.1 | NP_001102052.1 | 1 - 2 | 116 | 86 | |
XM_008761859.1 | XP_008760081.1 | 1 - 2 | 305 | 86 | ||
GenBank mRNA | BC168756.1 | - | 1 - 2 | 111 | 6887254 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
