Assay Details
| Gene Symbol: LOC679894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250641.2 | XP_006250703.1 | 2 - 3 | 5549 | 96 | |
| XM_006250642.2 | XP_006250704.1 | 2 - 3 | 5549 | 96 | ||
| XM_006250643.2 | XP_006250705.1 | 2 - 3 | 5549 | 96 | ||
| XM_008770063.1 | XP_008768285.1 | 2 - 3 | 5549 | 96 | ||
| XM_008770064.1 | XP_008768286.1 | 2 - 3 | 5549 | 96 | ||
| XM_008770065.1 | XP_008768287.1 | 2 - 3 | 5549 | 96 | ||
| XM_008770068.1 | XP_008768290.1 | 2 - 3 | 5549 | 96 | ||
| XM_008770069.1 | XP_008768291.1 | 2 - 3 | 7553 | 96 | ||
More Information
Set Membership: |
|

LOC679894
Genomic Map