Assay Details
| Gene Symbol: Sh3bgrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173339.1 | NP_001166810.1 | 160 | 101 | ||
| GenBank mRNA | BC169023.1 | - | 68 | 9025124 | 101 | |
| EV766621.1 | - | 1 - 2 | 160 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC169023.1, NM_001173339.1 transcripts.
Set Membership: |
|

Sh3bgrl
Genomic Map