Assay Details
| Gene Symbol: Sh3bgrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173339.1 | NP_001166810.1 | 349 | 78 | ||
| GenBank mRNA | BC169023.1 | - | 257 | 9025124 | 78 | |
| EV766621.1 | - | 2 - 3 | 349 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3bgrl
Genomic Map