Assay Details
| Gene Symbol: Ltb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212507.2 | NP_997672.1 | 2 - 3 | 568 | 102 | |
| GenBank mRNA | BC088208.1 | - | 2 - 3 | 568 | 7381953 | 102 |
| Gene Symbol: LOC103694381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772776.1 | XP_008770998.1 | 2 - 3 | 863 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ltb
Genomic Map