Assay Details
| Gene Symbol: Nkain4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106550.1 | NP_001100020.1 | 3 - 4 | 380 | 62 | |
| XM_006235744.2 | XP_006235806.1 | 5 - 6 | 634 | 62 | ||
| XM_006235745.2 | XP_006235807.1 | 5 - 6 | 2516 | 62 | ||
| XM_008762496.1 | XP_008760718.1 | 6 - 7 | 983 | 62 | ||
| XM_008762497.1 | XP_008760719.1 | 6 - 7 | 1811 | 62 | ||
| XM_008762498.1 | XP_008760720.1 | 6 - 7 | 786 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nkain4
Genomic Map