Assay Details
| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271328.1 | NP_001258257.1 | 5 - 6 | 578 | 96 | |
| XM_006252795.2 | XP_006252857.1 | 5 - 6 | 1400 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsh2d
Genomic Map