Assay Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011918.1 | NP_001011918.1 | 14 - 15 | 1520 | 76 | |
| XM_008770980.1 | XP_008769202.1 | 14 - 15 | 1766 | 76 | ||
| GenBank mRNA | BC083812.1 | - | 14 - 15 | 1520 | 7110191 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis
biological_process
response to calcium ion
molecular_function
calcium ion binding
calcium-dependent phospholipid binding
phosphatidylethanolamine binding
MHC class II protein complex binding
S100 protein binding
poly(A) RNA binding
calcium-dependent protein binding
phagocytosis
biological_process
response to calcium ion
molecular_function
calcium ion binding
calcium-dependent phospholipid binding
phosphatidylethanolamine binding
MHC class II protein complex binding
S100 protein binding
poly(A) RNA binding
calcium-dependent protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P070756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153026 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P070756 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P070756 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P070756 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P480490 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25045 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84753 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23293 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75401 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10786187 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786182 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786188 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786186 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786179 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786184 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786181 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786191 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786185 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786183 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786177 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786178 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786180 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10786174 | - | - |
| Rat Genome 230 2.0 Array | 1388445_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388445_at | - | - |
| Affymetrix HT Rat Focus Array | 1388445_at | - | - |
| Affymetrix HT Rat Focus Array | 1388445_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600008292.rn.1 | TC1600001133.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388445_at | - | - |
| Rat Expression Set 230 | 1388445_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786174 | - | - |
| Rat Genome U34 Set | rc_AI232030_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22395681 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376793 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376036 | - | - |

Anxa11
Genomic Map