Assay Details
| Gene Symbol: Mogat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109436.2 | NP_001102906.2 | 4 - 5 | 680 | 82 | |
| GenBank mRNA | BC166995.1 | - | 4 - 5 | 680 | 9027036 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Mogat2
Genomic Map