Assay Details
| Gene Symbol: Nfatc2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007692.1 | NP_001007693.1 | 3 - 4 | 688 | 104 | |
| GenBank mRNA | BC079050.1 | - | 3 - 4 | 688 | 7115323 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc2ip
Genomic Map