Assay Details
| Gene Symbol: Abca15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106293.2 | NP_001099763.2 | 1 - 2 | 142 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106293.2 transcripts.
Set Membership: |
|

Abca15
Genomic Map