Assay Details
| Gene Symbol: Zbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235674.2 | XP_006235736.1 | 2 - 3 | 3881 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235674.2 transcripts.
Set Membership: |
|

Zbp1
Genomic Map