Assay Details
| Gene Symbol: Sema3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104633.1 | NP_001098103.1 | 17 - 18 | 2455 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132422 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P885377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65311 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P885377 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132422 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132422 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P885377 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132422 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P885377 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16137 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P885377 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41707 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80424 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78005 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22209 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29641 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10860511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860509 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10860499 | - | - |
| Rat Genome 230 2.0 Array | 1397335_at | - | - |
| Rat Genome 230 2.0 Array | 1397335_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400016366.rn.1 | TC0400002304.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400016367.rn.1 | TC0400002304.rn.1 | RTA TC |
| Rat Expression Set 230 | 1397335_PM_at | - | - |
| Rat Expression Set 230 | 1397335_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860499 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21555136 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353557 | - | - |

Sema3d
Genomic Map