Assay Details
| Gene Symbol: Gngt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135777.1 | NP_001129249.1 | 1 - 2 | 134 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001135777.1 transcripts.
Set Membership: |
|

Gngt1
Genomic Map