Assay Details
| Gene Symbol: Snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106163.1 | NP_001099633.1 | 1 - 2 | 131 | 68 | |
| XM_008771943.1 | XP_008770165.1 | 1 - 2 | 146 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001106163.1,XM_008771943.1 transcripts.
This assay is in the 5'UTR region of the NM_001106163.1,XM_008771943.1 transcripts.
Set Membership: |
|

Snrpd1
Genomic Map