Assay Details
| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106428.2 | NP_001099898.2 | 4 - 5 | 889 | 62 | |
| XM_006232297.2 | XP_006232359.1 | 4 - 5 | 976 | 62 | ||
| XM_006232298.2 | XP_006232360.1 | 4 - 5 | 390 | 62 | ||
| GenBank mRNA | BC168157.1 | - | 4 - 5 | 889 | 8363916 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1049055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62165 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1049055 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1049055 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1049055 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1049055 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13468 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10815119 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815104 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815105 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815103 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815106 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815100 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815102 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815116 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10815099 | - | - |
| Rat Genome 230 2.0 Array | 1397739_at | - | - |
| Rat Genome 230 2.0 Array | 1397739_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378002_at | - | - |
| Rat Genome 230 2.0 Array | 1378002_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378002_at | - | - |
| Affymetrix HT Rat Focus Array | 1378002_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200006624.rn.1 | TC0200001045.rn.1 | RTA TC |
| Rat Expression Set 230 | 1378002_PM_at | - | - |
| Rat Expression Set 230 | 1378002_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815099 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20726889 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365783 | - | - |

Hspa4l
Genomic Map