Assay Details
| Gene Symbol: Prr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255942.1 | XP_006256004.1 | 1 - 2 | 128 | 148 | |
| GenBank mRNA | BC098636.1 | - | 1 - 2 | 138 | 148 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006255942.1 transcripts.
Set Membership: |
|

Prr3
Genomic Map