Assay Details
| Gene Symbol: Prr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212544.2 | NP_997709.1 | 1 - 2 | 140 | 121 | |
| XM_006255943.2 | XP_006256005.1 | 1 - 2 | 197 | 121 | ||
| GenBank mRNA | BC126073.1 | - | 1 - 2 | 140 | 7311139 | 121 |
More Information
Updated Mapping Information
Set Membership: |
|

Prr3
Genomic Map