Assay Details
| Gene Symbol: Myoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109097.1 | NP_001102567.1 | 3 - 4 | 330 | 73 | |
| XM_006251655.2 | XP_006251717.1 | 3 - 4 | 377 | 73 | ||
| GenBank mRNA | FQ214748.1 | - | 3 - 4 | 330 | 73 | |
| FQ214808.1 | - | 3 - 4 | 330 | 73 | ||
| FQ214853.1 | - | 3 - 4 | 313 | 73 | ||
| FQ215077.1 | - | 332 | 73 | |||
| FQ215190.1 | - | 3 - 4 | 332 | 73 | ||
| FQ215360.1 | - | 3 - 4 | 336 | 73 | ||
| FQ215382.1 | - | 3 - 4 | 332 | 73 | ||
| FQ215728.1 | - | 3 - 4 | 332 | 73 | ||
| FQ215901.1 | - | 3 - 4 | 334 | 73 | ||
| FQ215918.1 | - | 3 - 4 | 330 | 73 | ||
| FQ216019.1 | - | 3 - 4 | 370 | 73 | ||
| FQ216101.1 | - | 3 - 4 | 332 | 73 | ||
| FQ216115.1 | - | 3 - 4 | 331 | 73 | ||
| FQ216248.1 | - | 3 - 4 | 331 | 73 | ||
| FQ216396.1 | - | 3 - 4 | 314 | 73 | ||
| FQ216753.1 | - | 3 - 4 | 314 | 73 | ||
| FQ216842.1 | - | 3 - 4 | 314 | 73 | ||
| FQ217380.1 | - | 352 | 73 | |||
| FQ224894.1 | - | 3 - 4 | 334 | 73 | ||
| FQ224977.1 | - | 3 - 4 | 330 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myoz1
Genomic Map