Assay Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227796.2 | XP_006227858.1 | 1 - 2 | 233 | 66 | |
| XM_006227797.2 | XP_006227859.1 | 1 - 2 | 233 | 66 | ||
More Information
Set Membership: |
|

Zdhhc11
Genomic Map