Assay Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039342.2 | NP_001034431.2 | 8 - 9 | 993 | 83 | |
| XM_006227796.2 | XP_006227858.1 | 7 - 8 | 1005 | 83 | ||
| XM_006227797.2 | XP_006227859.1 | 7 - 8 | 954 | 83 | ||
| GenBank mRNA | AY886527.1 | - | 8 - 9 | 1083 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc11
Genomic Map